missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Blood Agar (TSA with Sheep Blood) Medium

Grow a wide variety of microorganisms on this general purpose non-selective medium that includes sheep blood to promote hemolysis.

Brand:  Thermo Scientific™ PB5012A

17.29 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10362223

  • £38.55 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Grow a wide variety of fastidious organisms on Thermo Scientific™ Blood Agar (TSA with Sheep Blood), a general purpose non-selective medium with the addition of sheep blood in the formulation to promote the presence and visualization of haemolytic reactions so it can be used as a diagnostic tool.

Tryptone Soya Agar (casein soya bean digest agar) with defibrinated sheep blood is suitable for the cultivation of fastidious aerobic and anaerobic bacteria, the latter of which can be grown either in deep cultures or by incubation under anaerobic conditions. The combination of casein digest and papaic digest of soybean meal leads to optimal growth due to synergy between the protein supply of casein and the carbohydrate supply of soybeans. Sterile defibrinated sheep blood used to enrich the medium produces the hemolysis characteristics of different bacteria, such as streptococci, listeria, hemolytic Staphylococcus, Escherichia coli and Pseudomonas.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Traffic red, opaque
90mm Monoplate
Blood Agar
17g ± 5%
Flow Wrapped - Single Layer
2-12°C, dark
Germany
Agar
TSA with Sheep Blood
10/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6254985 Certificate TypeCertificate of Analysis Date07/04/2025 Catalog NumberPB5012A
Lot Number6254984 Certificate TypeCertificate of Analysis Date07/04/2025 Catalog NumberPB5012A
Lot Number6254983 Certificate TypeCertificate of Analysis Date07/04/2025 Catalog NumberPB5012A
Lot Number6251998 Certificate TypeCertificate of Analysis Date07/04/2025 Catalog NumberPB5012A
Lot Number6251996 Certificate TypeCertificate of Analysis Date01/04/2025 Catalog NumberPB5012A
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Blood Agar (TSA with Sheep Blood) Medium >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.