missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Blotting Paper, Grade BF 3, Sheets
Sartorius Stedim Biotech™ Blotting Paper, Grade BF 3, Sheets are highly pure and ideal for blotting chromatography.
Brand: Sartorius Stedim Biotech™ FT-2-520-135155N
82.60 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- This highly pure paper grade is not only ideal for blotting and chromatography, but also for a wide range of absorption applications like those common in the life sciences and diagnostics.
- Made of high-purity cotton linters for uniform buffer flow and resulting blots.
- No additives to avoid any interference during transfer.
- BF 3 grade is ideal for increasing and maintaining the transport of liquid from the buffer and as buffer reservoir in capillary and semidry blotting methods.

Specifications
Capillary rise: 130mm/10 min., Weight: 330 g/m2, 100/Pk | |
0.76 mm | |
BF 3 | |
135 mm |
135 x 155 mm | |
Sheet | |
155 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sartorius Stedim Biotech™ Blotting Paper, Grade BF 3, Sheets > 135 x 155mm
Spot an opportunity for improvement?Share a Content Correction