Learn More
Manutan Blue medium duty steel shelving starter bay, 2000mm x 1250mm x 500mm
Brand: Manutan A360325
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Description
Economic multipurpose shelving. All steel for durability with easy 'click-in' system.
- Five shelves
- Blue uprights
- Shelf capacity UDL: 110kg
- Dimensions (H x W x D): 2000mm x 1250mm x 500mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.