missing translation for 'onlineSavingsMsg'
Learn More

Kautex™ Blue Polypropylene Screw Closures

Provides tamper evidence with pull-off ring and conical seal. Kautex™ Blue Polypropylene Screw Closures are ribbed for easy grip opening.

Brand:  Kautex™ 2000083951

 View more versions of this product

Product Code. 15961441

  • £54.00 / Pack of 100
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Leakproof conical seal
  • Blue color for content identification
  • Polypropylene construction for strength and durability
  • Available for bottles with capacities of 10 to 2500 mL
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blue
Polypropylene
10 to 1000 mL Bottle
Pull-off Ring and Conical Seal
Screw Cap
32 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.