missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Blue Single-Use Tweezers
GREENER_CHOICEBIOPHARMA

Tweezers in a unique shade of blue that does not occur often in foodstuffs, making them easy to find and visually identify.

£73.20 - £78.95

Specifications

No. per Pack 100 (packaged individually)
Length (English) 5.12 in.
Material Polystyrene
Length (Metric) 130 mm
Product Type Forceps
View More Specs

Products 3
Product Code Brand Tip Style Price Quantity & Availability  
Product Code Brand Tip Style Price Quantity & Availability  
16565210
GREENER_CHOICEBIOPHARMA
View Documents
Buerkle™
5378-1050
Angled, Pointed
£73.20
Pack of 100
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
 
16575210
GREENER_CHOICEBIOPHARMA
View Documents
Buerkle™
5378-1051
Pointed
£78.95
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
16585210
GREENER_CHOICEBIOPHARMA
View Documents
Buerkle™
5378-1052
Broad
£75.15
Pack of 100
Estimated Shipment: 08-05-2025
to see stock.
Add to basket
 
Description

Description

  • The use of blue tools thus helps to meet the requirements of the HACCP hazard analysis and assessment in reference to foreign objects
  • This is an important criterion, especially in food processing and production and in the animal feed and pharmaceutical industry
  • The disposable forceps are suitable for precise lab work and are ideal for grasping, lifting and moving small objects, for example fine tissue, small items or solid samples
  • The fluted handles ensure optimum and secure handling, as well as precise work
  • The light plastic forceps for single use allow for fatigue-free work
  • The clean room-manufactured lab assistants meet the highest standards and prevent cross-contamination or impurities in the sample being examined
  • The lab tweezers are suitable for chemical, pharmaceutical or cosmetic applications as well as the food industry and scientific uses
  • Individually packaged
  • For single use
  • Clean room manufactured (cleanroom class 7)
  • Complies with EU food requirements and FDA regulations
  • Pointed/angled tip: The disposable forceps have fine, smooth, curved gripping tips. Different materials can be held and processed reliably with the pointed ends
  • Pointed tip: The anatomical forceps have a straight tip, fluted on the inside. This makes it possible to grip sensitive materials reliably
  • Broad tip: The disposable forceps have ends with a broad tip. The broad tip is smooth and blunt. It prevents the material from being damaged when it is gripped
Specifications

Specifications

100 (packaged individually)
Polystyrene
Forceps
Standard
5.12 in.
130 mm
Sterile
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buerkle™ Blue Single-Use Tweezers

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.