Learn More
Buerkle™ Blue Single-Use Tweezers



Tweezers in a unique shade of blue that does not occur often in foodstuffs, making them easy to find and visually identify.
Brand: Buerkle™ 5378-1052
59.18 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- The use of blue tools thus helps to meet the requirements of the HACCP hazard analysis and assessment in reference to foreign objects
- This is an important criterion, especially in food processing and production and in the animal feed and pharmaceutical industry
- The disposable forceps are suitable for precise lab work and are ideal for grasping, lifting and moving small objects, for example fine tissue, small items or solid samples
- The fluted handles ensure optimum and secure handling, as well as precise work
- The light plastic forceps for single use allow for fatigue-free work
- The clean room-manufactured lab assistants meet the highest standards and prevent cross-contamination or impurities in the sample being examined
- The lab tweezers are suitable for chemical, pharmaceutical or cosmetic applications as well as the food industry and scientific uses
- Individually packaged
- For single use
- Clean room manufactured (cleanroom class 7)
- Complies with EU food requirements and FDA regulations
- Pointed/angled tip: The disposable forceps have fine, smooth, curved gripping tips. Different materials can be held and processed reliably with the pointed ends
- Pointed tip: The anatomical forceps have a straight tip, fluted on the inside. This makes it possible to grip sensitive materials reliably
- Broad tip: The disposable forceps have ends with a broad tip. The broad tip is smooth and blunt. It prevents the material from being damaged when it is gripped
Specifications
Broad | |
5.12 in. | |
130 mm | |
Sterile |
100 (packaged individually) | |
Polystyrene | |
Forceps | |
Standard |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.