Learn More
Brady™ BMP51 Label Printer Kit, EU
Multi-featured portable label maker for quickly making labels on the go. Brady™ BMP51 Label Printer Kit includes a built-in cutter and uses easy, top-loading cartridges. EU plug.
Brand: Brady™ 710896
Code : NEW
Additional Details : Weight : 5.55100kg Transport : UN number : 3481 Chem class : 9
Description
- 300 DPI print head.
- Prints labels up to 38.1mm wide.
- Prints on continuous and die-cut materials.
- Includes built-in cutter, easy top-loading cartridges, options for battery power
- Full QWERTY keyboard
Specifications
Label Printer | |
Thermal Transfer | |
90 mm | |
38.1 mm | |
QWERTY | |
Danish, Dutch, English, Finnish, French, German, Italian, Norwegian (Bokmål), Portuguese, Spanish, Swedish | |
25.4 mm/sec. | |
LabelMark | |
1.4 kg | |
1 Ea. | |
Lithium ion battery and AC power |
Monocolor LCD | |
115 mm | |
225 mm | |
Monochrome | |
This printer ships in a hard case with a Rechargeable Lithium Ion Battery, USB Cable, AC adaptor with power cord, Product CD, Brady Printer Drivers CD, Quick Start Guide, a Bluetooth network card, LabelMark Standard Labelling Software and an MC1-1000-422 | |
48 MB - up to 25,000 labels | |
300 dpi | |
USB, Bluetooth. Optional: Ethernet, WiFi | |
Electrical, datacom, facility/safety, product and panel manufacturing, and laboratory labelling applications | |
2 Years |