Learn More
Tocris Bioscience™ BMS 566419
Inosine monophosphatase dehydrogenase (IMPDH) inhibitor
Brand: Tocris Bioscience™ 5492/10
114.03 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
BMS 566419 is a potent inosine monophosphate dehydrogenase (IMPDH) inhibitor (IC50 = 17 nM). Inhibits proliferation of human T-lymphoblasts and PBMCs in vitro. Reduces paw swelling in a rat adjuvant arthritis model. Orally available.Specifications
N-[1-[6-(4-Ethyl-1-piperazinyl)-3-pyridinyl]-1-methylethyl]-2-fluoro-9,10-dihydro-9-oxo-3-acridinecarboxamide | |
10 mg | |
C28H30FN5O2 | |
>98% |
566161-24-8 | |
Dehydrogenase Inhibitors | |
487.57 |