missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BMS 566419
Inosine monophosphatase dehydrogenase (IMPDH) inhibitor
£142.54 - £490.27
Specifications
Chemical Name or Material | N-[1-[6-(4-Ethyl-1-piperazinyl)-3-pyridinyl]-1-methylethyl]-2-fluoro-9,10-dihydro-9-oxo-3-acridinecarboxamide |
---|---|
CAS | 566161-24-8 |
Target | Dehydrogenase Inhibitors |
Molecular Formula | C28H30FN5O2 |
Formula Weight | 487.57 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18746148
|
Tocris Bioscience™
5492/50 |
50 mg |
£490.27
50mg |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18740278
|
Tocris Bioscience™
5492/10 |
10 mg |
£142.54
10mg |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Potent inosine monophosphate dehydrogenase (IMPDH) inhibitor (IC50 = 17 nM). Inhibits proliferation of human T-lymphoblasts and PBMCs in vitro. Reduces paw swelling in a rat adjuvant arthritis model. Orally available.Specifications
N-[1-[6-(4-Ethyl-1-piperazinyl)-3-pyridinyl]-1-methylethyl]-2-fluoro-9,10-dihydro-9-oxo-3-acridinecarboxamide | |
Dehydrogenase Inhibitors | |
487.57 |
566161-24-8 | |
C28H30FN5O2 | |
>98% |