missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Boc-Val-Leu-Gly-Arg-pNA Acetate Salt
Boc-Val-Leu-Gly-Arg-pNA is a chromogenic substrate for horseshoe crab clotting enzyme for endotoxins detection.
Brand: MP Biomedicals™ 08613082
Additional Details : Weight : 0.00010kg
Description
- Boc-VLGR-pNA acetate salt (sequence Boc-Val-Leu-Gly-Arg-pNA), is a chromogenic substrate for horseshoe crab clotting enzyme
- It has been widely used in detection assays for bacterial endotoxins
Specifications
-20°C | |
Endotoxin detection | |
100 mg |
Powder | |
663.78 |