missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Boc-Val-Leu-Gly-Arg-pNA Acetate Salt
Boc-Val-Leu-Gly-Arg-pNA is a chromogenic substrate for horseshoe crab clotting enzyme for endotoxins detection.
£192.92 - £576.64
Specifications
Content And Storage | -20°C |
---|---|
Format | Powder |
For Use With (Application) | Endotoxin detection |
Molecular Weight (g/mol) | 663.78 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16369845
|
MP Biomedicals™
08613081 |
25 mg |
£192.92
25mg |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
16379845
|
MP Biomedicals™
08613082 |
100 mg |
£576.64
100mg |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
Description
- Boc-VLGR-pNA acetate salt (sequence Boc-Val-Leu-Gly-Arg-pNA), is a chromogenic substrate for horseshoe crab clotting enzyme
- It has been widely used in detection assays for bacterial endotoxins
Specifications
-20°C | |
Powder | |
Endotoxin detection | |
663.78 |