missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ FEP Dropping Funnels with PTFE stopecock

Designed for easy locking and unlocking of the ground joint. BOLA dropping funnels are suited for a variety of applications.

Brand:  Bohlender™ B285-02

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10620752

  • £526.82 / Each
Estimated Shipment: 14-08-2024
to see stock.
Add to basket

Description

Description

  • Transparent FEP Dropping funnel
  • Fine adjustment stopcock with cone size 29 made of PTFE
  • Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint
  • Compliance

    FDA

Specifications

Specifications

250 mL
FDA
Transparent
+110°C
Dropping funnel with stopcock
Plastic
Yes
29
Polytetrafluoroethylene (PTFE)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers