missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bohlender™ BOLA™ Flexible Tubing EX
Flexible tubing with cylindrical tubing ends for a better connection.
Brand: Bohlender™ S1824-57
This item is not returnable.
View return policy
Description
- Conductive corrugated tubing
- Circular corrugations around the longitudinal axis
- Cylindrical tubing ends with a length of 40 mm can for example be connected directly to fittings, stopcocks or hose connectors
- Together with BOLA™ Laboratory Screw Joints EX, the connection is conductive, absolutely tight and suitable for vacuum
Specifications
6 mm | |
8 mm | |
1 m | |
10 mm | |
Cylindrical | |
11 bar |
PFA-EX | |
3.3 ft. | |
BOLA™ Laboratory Screw Joints EX | |
0.4 in. | |
18 mm bend radius |