Learn More
Thermo Scientific™ Bond-Breaker™ TCEP Solution, Neutral pH
Efficiently reduce protein disulfide bonds with this stable, odorless, 0.5M solution of TCEP reducing agent that is ideal for making reducing SDS-PAGE sample buffers.
Brand: Thermo Scientific™ 77720
196.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Code : PN
Additional Details : CAS Number : 51805-45-9 Weight : 0.45360kg Transport : UN number : 3265 Chem class : 8 Pack group : III
Description
Thermo Scientific™ Bond-Breaker TCEP Solution, Neutral pH is a stable, 0.5M solution of the thiol-free, phosphine-based TCEP compound, useful as a 10X stock for addition to SDS-PAGE sample loading buffers to reduce protein disulfide bonds.
Highlights:
- Ready-to-use, odorless, stable and neutral 0.5M TCEP solution
- Eliminates TCEP·HCl stock solution preparation and neutralization prior to SDS-PAGE gel loading
- Convenient, time-saving format for TCEP reductions
- Neutral pH minimizes possibility of amide bond cleavage during reduction
- Room temperature-stable, saves valuable refrigerator space
- Contributes to more pleasant, safer laboratory environment
Specifications
Store at room temperature. | |
Liquid | |
Disulfide Reducing Reagent |
Bond-Breaker™ | |
5 mL |
Safety and Handling
- TCEP Solution
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : Propanoic acid, 3, 3', 3''-phosphinidynetris-, hydrochloride
For Research Use Only. Not for use in diagnostic procedures.