missing translation for 'onlineSavingsMsg'
Learn More

Restek™ Bonded Reversed Phase Resprep SPE Cartridges

Hydrophobic (nonpolar) silica-based adsorbents; used to extract hydrophobic analytes from polar matrices.

Brand:  Restek™ 24051

Encompass_Preferred

 View more versions of this product

Product Code. 13189942

  • £211.75 / Pack of 30
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured using high density polypropylene
  • May be processed by any of these techniques: positive pressure, sidearm flask, centrifuge or vacuum manifold

Specifications

Specifications

Hydrophobic (non-Polar)
Reversed Bonded
SPE Cartridge
Yes
30/Pk.
Silica
C18 (high load)
6 mL, 1 g
HDPE
6 mL, 1,000 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers