missing translation for 'onlineSavingsMsg'
Learn More
Borane-d{3}, 1M in tetrahydrofuran, Thermo Scientific Chemicals Product Code.: 15412997

Borane-d{3}, 1M in tetrahydrofuran, Thermo Scientific Chemicals

Product Code. 15412997
250 mL
Click to view available options
Quantity:
50 mL
250 mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15412997

Brand: Thermo Scientific Alfa Aesar 089121.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13763-62-7
Molecular Formula BD3
Molecular Weight (g/mol) 16.85
MDL Number MFCD00064892
InChI Key UORVGPXVDQYIDP-FIBGUPNXSA-N
Synonym borane-d3, borane-d3, 1m in tetrahydrofuran, borane-d3, 1m in tetrahydrofuran, packaged under argon in resealable chemseal bottlesShow MoreShow Less
PubChem CID 11062299
SMILES [2H]B([2H])[2H]

Specifications

Color Colorless
Physical Form Liquid
Quantity 250 mL
UN Number UN3148
Flash Point -21°C (-6°F)
IUPAC Name (²H₃)borane
Formula Weight 16.84 as BD3
Concentration or Composition (by Analyte or Components) 1M in tetrahydrofuran
Odor Ether-like
Sensitivity Air & Moisture Sensitive
Density 0.876g/mL
Chemical Name or Material Borane-d_3
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU18I009 Certificate TypeCertificate of Analysis Date22/09/2022 Catalog Number089121.JS,089121.AK,089121.JP,089121.AD
Lot NumberD01W045 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number089121.JS,089121.AK,089121.JP,089121.AD
Lot NumberQ26C036 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number089121.JS,089121.AK,089121.JP,089121.AD
Lot NumberF15U004 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number089121.JS,089121.AK,089121.JP,089121.AD
Lot NumberT18A025 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number089121.JS,089121.AK,089121.JP,089121.AD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Borane-d{3}, 1M in tetrahydrofuran, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.