missing translation for 'onlineSavingsMsg'
Learn More

Borane-pyridine complex, 95%, Thermo Scientific Chemicals

£38.10 - £127.64

Chemical Identifiers

CAS 110-51-0
Molecular Formula C5H8BN
Molecular Weight (g/mol) 92.94
MDL Number MFCD00012435
InChI Key LPGWNCNRGQANGC-UHFFFAOYSA-N
Synonym Pyridine-borane
IUPAC Name (pyridin-1-ium-1-yl)boranuide
SMILES [BH3-][N+]1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11426790
View Documents
Thermo Scientific Alfa Aesar
L13178.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11436790
View Documents
Thermo Scientific Alfa Aesar
L13178.18
50 g
£127.64
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Borane-pyridine complex is used for synthesis of α-alkyl-β-hydroxy esters and preparation of Igs conjugated with viral peptide epitopes. It is used as reducing agent for high performance liquid chromatography analysis of oligosaccharides. It is also used as modifier in decant oil affecting its carbonization to mesophase pitch.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Borane-pyridine complex is used for synthesis of α-alkyl-β-hydroxy esters and preparation of Igs conjugated with viral peptide epitopes. It is used as reducing agent for high performance liquid chromatography analysis of oligosaccharides. It is also used as modifier in decant oil affecting its carbonization to mesophase pitch.

Solubility
It hydrolyzes slowly in water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and heat.
Specifications

Chemical Identifiers

110-51-0
92.94
LPGWNCNRGQANGC-UHFFFAOYSA-N
(pyridin-1-ium-1-yl)boranuide
C5H8BN
MFCD00012435
Pyridine-borane
[BH3-][N+]1=CC=CC=C1

Specifications

110-51-0
0.92
73°C (163°F)
C5H8BN
MFCD00012435
3674913
Pyridine-borane
LPGWNCNRGQANGC-UHFFFAOYSA-N
(pyridin-1-ium-1-yl)boranuide
92.93
Borane-pyridine complex
9°C to 11°C
65°C (1 mmHg)
Unpleasant
1.532
UN2810
Moisture sensitive
It hydrolyzes slowly in water.
[BH3-][N+]1=CC=CC=C1
92.94
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H310-H330-H227-H315-H319-H335
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P210-P235-P260-P262-P264b-P270-P271-P281-P284-P301+P310-P302+P350-P304+P340-P308+P313-P310-P330-P361-P363-P370+P378q-P501c

H227-H301-H310+H330-H351

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 203-773-4

missing translation for 'rtecsNumber' : US3675000

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only