Learn More
Borane-tetrahydrofuran complex, 1M solution in THF, Stabilized, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 175088000
242.64 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
Specifications
Borane-tetrahydrofuran complex | |
1M solution in tetrahydrofuran, stabilized | |
C4H11BO | |
0.96 to 1.08M | |
0.8760g/mL | |
borane-tetrahydrofuran complex, tetrahydrofuran borane, bh3.thf, borane tetrahydrofuran complex solution, borane-d3-thf complex solution, borane-tetrahydrofuran, unii-5ear4err1l, oxolane borane, boron; oxolane, borane thf | |
RMCYTHFAWCWRFA-UHFFFAOYSA-N | |
oxolane borane | |
11062302 | |
AcroSeal™ Glass bottle | |
Stabilized | |
−22°C |
Colorless | |
14044-65-6, 109-99-9 | |
MFCD00012429 | |
01,199; 02,106; 03,76; 04,124; 05,184; 06,161; 07,89; 12,65; 17,101 | |
15, 1336 | |
Solubility in water: reacts. | |
B.C1CCOC1 | |
85.94 | |
85.94 | |
0.876 | |
800 mL | |
Liquid |
Safety and Handling
- Borane-tetrahydrofuran complex, 1M solution in THF, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0465002 | Certificate of Analysis | 31/01/2025 | 175088000,175081000 |
A0455203 | Certificate of Analysis | 31/01/2025 | 175088000,175081000 |
A0465002 | Certificate of Analysis | 31/07/2024 | 175088000,175081000 |
A0455203 | Certificate of Analysis | 25/01/2024 | 175088000,175081000 |
A0451293 | Certificate of Analysis | 25/01/2024 | 175088000,175081000 |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
0Hours
18Minutes
43Seconds
23 -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
31Hours
18Minutes
43Seconds
23New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only