missing translation for 'onlineSavingsMsg'
Learn More
Boric acid, 99.6%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10435281

Boric acid, 99.6%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10435281
2 kg, Plastic bottle
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10435281

Brand: Thermo Scientific Acros 423480020

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-35-3
Molecular Formula BH3O3
Molecular Weight (g/mol) 61.83
MDL Number MFCD00011337
InChI Key KGBXLFKZBHKPEV-UHFFFAOYSA-N
Synonym orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufeShow MoreShow Less
PubChem CID 7628
ChEBI CHEBI:33118
IUPAC Name boric acid
SMILES OB(O)O

Specifications

Color Brown-Red
Heavy Metals (as Pb) 10ppm max.
Non_volatile Matter 0.05 % max. (with MeOH)
Packaging Plastic bottle
Assay Percent Range 99.6%
Quantity 2 kg
Linear Formula H3BO3
Physical Form Crystalline powder
Fieser 01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,70
Merck Index 15, 1339
Formula Weight 61.83
Percent Purity ≥99.5% (Acid)
Grade ACS Reagent
Chemical Name or Material Boric acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468204 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number423480020,423485000
Lot NumberA0468208 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number423480020,423485000
Lot NumberB0156309 Certificate TypeCertificate of Analysis Date06/12/2024 Catalog Number423480020,423485000
Lot NumberA0463557 Certificate TypeCertificate of Analysis Date05/06/2024 Catalog Number423480020,423485000
Lot NumberA0463477 Certificate TypeCertificate of Analysis Date03/06/2024 Catalog Number423480020,423485000
compliance-icons
Product Identifier
  • Boric acid
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boric acid, 99.6%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.