Learn More
Boric acid, 99.6%, ACS reagent, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Brown-Red |
Heavy Metals (as Pb) | 10ppm max. |
Non_volatile Matter | 0.05 % max. (with MeOH) |
Packaging | Plastic bottle |
Assay Percent Range | 99.6% |
Quantity | 500 g |
Linear Formula | H3BO3 |
Physical Form | Crystalline powder |
Fieser | 01,63; 02,32; 03,29; 04,41; 05,48; 09,59; 11,70 |
Merck Index | 15, 1339 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0468204 | Certificate of Analysis | 25/01/2025 | 423480020,423485000 |
A0468208 | Certificate of Analysis | 25/01/2025 | 423480020,423485000 |
B0156309 | Certificate of Analysis | 06/12/2024 | 423480020,423485000 |
A0463557 | Certificate of Analysis | 05/06/2024 | 423480020,423485000 |
A0463477 | Certificate of Analysis | 03/06/2024 | 423480020,423485000 |
Safety and Handling

- Boric acid
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : boric acid
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
57Hours
14Minutes
9Seconds
36New Offer Save Up to 70%
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.