missing translation for 'onlineSavingsMsg'
Learn More
Boron oxide, 98%, Thermo Scientific Chemicals Product Code.: 11409223

Boron oxide, 98%, Thermo Scientific Chemicals

Product Code. 11409223
2500 g
Click to view available options
Quantity:
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11409223

Brand: Thermo Scientific Alfa Aesar A11707.0E

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants.

Solubility
Soluble in water. Slightly soluble in methanol.

Notes
Hygroscopic and moisture sensitive. Incompatible with acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1303-86-2
Molecular Formula B2O3
Molecular Weight (g/mol) 69.62
MDL Number MFCD00011315
InChI Key JKWMSGQKBLHBQQ-UHFFFAOYSA-N
Synonym boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g
PubChem CID 518682
ChEBI CHEBI:30163
IUPAC Name oxo(oxoboranyloxy)borane
SMILES O=BOB=O

Specifications

Boiling Point 1860°C
Melting Point ∽450°C
Quantity 2500 g
Merck Index 14,1337
Solubility Information Soluble in water. Slightly soluble in methanol.
Formula Weight 69.62
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 2.46 +0.01
Chemical Name or Material Boron oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW13K026 Certificate TypeCertificate of Analysis Date18/03/2025 Catalog NumberA11707.0E,A11707.36
Lot NumberQ03J013 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA11707.0E,A11707.36
Lot NumberW12I022 Certificate TypeCertificate of Analysis Date04/10/2023 Catalog NumberA11707.0E,A11707.36
Lot NumberY16G050 Certificate TypeCertificate of Analysis Date14/06/2022 Catalog NumberA11707.0E,A11707.36
Lot Number10239659 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA11707.0E,A11707.36
compliance-icons
Product Identifier
  • Boron oxide
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Boron oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boron oxide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.