Learn More
Boron oxide, 98%, Thermo Scientific Chemicals
£48.80 - £191.00
Chemical Identifiers
CAS | 1303-86-2 |
---|---|
Molecular Formula | B2O3 |
Molecular Weight (g/mol) | 69.62 |
MDL Number | MFCD00011315 |
InChI Key | JKWMSGQKBLHBQQ-UHFFFAOYSA-N |
Synonym | boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g |
PubChem CID | 518682 |
ChEBI | CHEBI:30163 |
IUPAC Name | oxo(oxoboranyloxy)borane |
SMILES | O=BOB=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499213
|
Thermo Scientific Alfa Aesar
A11707.36 |
500 g |
£48.80
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409223
|
Thermo Scientific Alfa Aesar
A11707.0E |
2500 g |
£191.00
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Boron oxide is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBoron oxide is used as a flux, an acid catalyst and a chemical intermediate in production of boron halides, esters, carbide, nitride and metallic borides. Boron oxide has shown a promising effect on glycerol hydrogenolysis to 1,2-propanediol. It is also an essential material for the preparation of cordierite ceramics from talc. It is actively involved in the production of borosilicate glass. It is an additive used in high-temperature lubricants.
Solubility
Soluble in water. Slightly soluble in methanol.
Notes
Hygroscopic and moisture sensitive. Incompatible with acids and strong oxidizing agents.
Chemical Identifiers
1303-86-2 | |
69.62 | |
JKWMSGQKBLHBQQ-UHFFFAOYSA-N | |
518682 | |
oxo(oxoboranyloxy)borane |
B2O3 | |
MFCD00011315 | |
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g | |
CHEBI:30163 | |
O=BOB=O |
Specifications
1860°C | |
1303-86-2 | |
MFCD00011315 | |
boron trioxide, boric oxide, boric anhydride, boric acid anhydride, oxo oxoboranyloxy borane, boronoxide, diboron trioxide, boron sesquioxide, boron oxide, boron oxide 500g | |
JKWMSGQKBLHBQQ-UHFFFAOYSA-N | |
oxo(oxoboranyloxy)borane | |
518682 | |
69.62 | |
Odorless | |
2.46 +0.01 |
∽450°C | |
B2O3 | |
14,1337 | |
Soluble in water. Slightly soluble in methanol. | |
O=BOB=O | |
69.62 | |
CHEBI:30163 | |
99% | |
Hygroscopic | |
Boron oxide |
Safety and Handling
GHS H Statement
H360
May damage fertility or the unborn child.
P201-P202-P281-P308+P313-P501c
H360FD
missing translation for 'einecsNumber' : 215-125-8
missing translation for 'rtecsNumber' : ED7900000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only