missing translation for 'onlineSavingsMsg'
Learn More

Boron standard solution, for AAS, 1mg/mL B in water, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 195861000

18.43 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.



 View more versions of this product

Product Code. 10154260

  • £19.40 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Boron standard solution
Plastic bottle
B
Solution
997 to 1003 μg/mL of B in water (20°C)
[B]
Soluble in water
5462311
10.81
7440-42-8
100 mL
MFCD00134034 MFCD00151272
14, 1343
ZOXJGFHDIHLPTG-UHFFFAOYSA-N
boron
10.81
CHEBI:27560
AAS
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Reproductive toxicity Category 1B

Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0450858 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number195865000,195861000
Lot NumberA0385901 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number195865000,195861000
Lot NumberA0441202 Certificate TypeCertificate of Analysis Date18/04/2022 Catalog Number195865000,195861000
Lot NumberA0437634 Certificate TypeCertificate of Analysis Date24/12/2021 Catalog Number195865000,195861000
Lot NumberA0431888 Certificate TypeCertificate of Analysis Date01/07/2021 Catalog Number195865000,195861000
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Boron standard solution, for AAS, 1mg/mL B in water, Thermo Scientific Chemicals > 100mL; Plastic bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only