Learn More
Sulfuric acid, 0.1N (0.05M) standard solution, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 124230010
30.64 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solution
Specifications
Sulfuric acid | |
0.1N (0.05M) Standard Solution | |
0.0998 to 0.1002N (20°C) | |
Plastic Bottle | |
H2O4S | |
Liquid | |
01,470; 05,633; 06,558; 07,347; 09,441 | |
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
OS(O)(=O)=O | |
98.07 | |
CHEBI:26836 | |
Pure |
Colorless | |
7664-93-9, 7732-18-5 | |
1.3g/mL | |
1 L | |
H2SO4 | |
MFCD00064589 | |
15, 9104 | |
QAOWNCQODCNURD-UHFFFAOYSA-N | |
sulfuric acid | |
1118 | |
98.07 |
Safety and Handling
- Sulfuric acid, 0.1N (0.05M) standard solution
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0464880 | Certificate of Analysis | 09/08/2024 | 124230025,124230010 |
A0464881 | Certificate of Analysis | 09/08/2024 | 124230025,124230010 |
B00Y5419 | Certificate of Analysis | 11/06/2024 | 124230025,124230010 |
A0458455 | Certificate of Analysis | 18/12/2023 | 124230025,124230010,124230250 |
B00V3206 | Certificate of Analysis | 14/11/2023 | 124230025,124230010,124230250 |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only