Learn More
Boron trifluoride dimethanol complex, 50-52 wt% BF3, Thermo Scientific Chemicals
£24.30 - £76.70
Chemical Identifiers
CAS | 2802-68-8, 7637-07-2, 67-56-1 |
---|---|
Molecular Formula | C2H8BF3O2 |
Molecular Weight (g/mol) | 131.89 |
MDL Number | MFCD00071635 |
InChI Key | MPRMFRXCVMCOMX-UHFFFAOYSA-N |
Synonym | boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution |
PubChem CID | 11062313 |
SMILES | CO.CO.FB(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10091990
|
Thermo Scientific Acros
158900500 |
50 g | Glass bottle |
£24.30
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10387570
|
Thermo Scientific Acros
158902500 |
250 g | Glass bottle |
£33.40
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10307530
|
Thermo Scientific Acros
158900010 |
1 kg | Glass bottle |
£76.70
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
2802-68-8, 7637-07-2, 67-56-1 | |
131.89 | |
MPRMFRXCVMCOMX-UHFFFAOYSA-N | |
11062313 |
C2H8BF3O2 | |
MFCD00071635 | |
boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution | |
CO.CO.FB(F)F |
Specifications
2802-68-8, 7637-07-2, 67-56-1 | |
Colorless to Yellow | |
59.0°C (4.0 mmHg) | |
C2H8BF3O2 | |
1.34 | |
1.222 | |
Solubility in water: hydrolysis. | |
CO.CO.FB(F)F | |
131.89 | |
131.89 | |
Liquid |
-20.0°C | |
1.2220g/mL | |
68°C | |
BF3·2CH3OH | |
MFCD00071635 | |
boron trifluoride-methanol solution, boron trifluoride-methanol complex, boron trifluoride methanol, boron trifluoride-methanol solution in methanol, bf3 methanol, .bf3 in methanol, boron fluoride methanol, bf3 meoh, borontrifluoride methanol, boron trifluoride solution | |
MPRMFRXCVMCOMX-UHFFFAOYSA-N | |
methanol;trifluoroborane | |
11062313 | |
≥50% (BF3) | |
Boron trifluoride dimethanol complex, 50-52 wt% BF3 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes damage to organs through prolonged or repeated exposure.<br/
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 220-543-9
missing translation for 'tsca' : TSCA
- Boron trifluoride dimethanol complex
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H370-Causes damage to organs.
- H372-Causes damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Methyl alcohol, Borate(1-), trifluoromethoxy-, (T-4)-, hydrogen, compound with methanol (1:1), Boron
RUO – Research Use Only