Learn More
BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper, Hexagonal Grip
Hollow with drip tip and hexagonal grip. BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper is constructed from Boro 3.3 to DIN 12252 standards.
Brand:  BRAND™ 144625
5.40 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10039970
  • £7.35 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Borosilicate glass constrcution (Boro 3.3. to DIN 12252)
  • Hollow
  • Hexagonal grip
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1/Pk.
Borosilicate Glass
Conical
NS 19/26
Ground Joint Stopper
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper, Hexagonal Grip > Size: NS 19/26