missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper, Hexagonal Grip
Hollow with drip tip and hexagonal grip. BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper is constructed from Boro 3.3 to DIN 12252 standards.
£4.90 - £26.85
Specifications
Material | Borosilicate Glass |
---|---|
Product Type | Ground Joint Stopper |
Shape | Conical |
Quantity | 1/Pk. |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
10253201
|
BRAND™
144635 |
NS 29/32 |
£8.05
Each |
In Stock
Log in to see stock. |
||||||
10039970
|
BRAND™
144625 |
NS 19/26 |
£7.35
Each |
In Stock
Log in to see stock. |
||||||
10781573
|
BRAND™
144615 |
NS 12/21 |
£4.90
Each |
In Stock
Log in to see stock. |
||||||
10719011
|
BRAND™
144620 |
NS 14/23 |
£6.85
Each |
In Stock
Log in to see stock. |
||||||
10408952
|
BRAND™
144630 |
NS 24/29 |
£10.85
Each |
In Stock
Log in to see stock. |
||||||
10613592
|
BRAND™
144645 |
NS 45/40 |
£26.85
Each |
In Stock
Log in to see stock. |
||||||
10141441
|
BRAND™
144605 |
NS 7/16 |
£4.90
Each |
In Stock
Log in to see stock. |
||||||
Description
- Borosilicate glass constrcution (Boro 3.3. to DIN 12252)
- Hollow
- Hexagonal grip
Specifications
Borosilicate Glass | |
Ground Joint Stopper | |
Conical | |
1/Pk. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper, Hexagonal Grip