Learn More
BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper, Hexagonal Grip
Hollow with drip tip and hexagonal grip. BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper is constructed from Boro 3.3 to DIN 12252 standards.
Brand:  BRAND™ 144630
6.49 GBP valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 10408952
  • £10.85 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Borosilicate glass constrcution (Boro 3.3. to DIN 12252)
  • Hollow
  • Hexagonal grip
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1/Pk.
NS 24/29
Ground Joint Stopper
NS 24/29
Borosilicate Glass
Conical
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BRAND™ BISTABIL™ Borosilicate Glass Conical Ground Joint Stopper, Hexagonal Grip > NS 24/29