Learn More
BRAND™ BLAUBRAND™ Borosilicate Glass Centrifuge Tube
Withstands RCF up to 700. Design and accuracy meet ASTM (American Society for Testing and Materials) specifications.
Brand:  BRAND™ 362138
 View more versions of this product
Product Code. 11884050
  • £142.00 / Pack of 2
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Withstands RCF up to 700.
  • Borosilicate glass construction (Boro 3.3)
  • Cylindrical with conical bottom
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 mL
160 mm
Conical
700 x g