Learn More
Cytiva Borosilicate Glass Chromatographic Tubes
Resistant to aqueous solutions and solvents used in liquid chromatography
Brand: Cytiva 19-5137-01
This item is not returnable.
View return policy
Description
Not resistant to acetone, ketones, chlorinated hydrocarbons, aliphatic esters, phenol, NaOH, > 10% HCl, > 5% acetic acid, or strong mineral acid.Specifications
For C 16/20 Columns | |
16 mm | |
14.5 psi |
Chromatography Tubes | |
Borosilicate Glass | |
60°C |