Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Conical Separating Funnels with Solid Glass Plug, Scale
Made of DURAN™ borosilicate glass. Lenz Laborglasinstrumente™ Borosilicate Glass Conical Separating Funnels with Solid Glass Plug, Scale are perfect for liquid-liquid extractions and separating chemical components of a mixture into immiscible solvent phases of different densities.
Brand:  Lenz Laborglasinstrumente™ 04002070
 View more versions of this product
Product Code. 11822672
  • £127.00 / Pack of 2
Estimated Shipment: 26-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS socket and PE stopper
  • with solid glass plug
  • with screwthread retaining device
  • with scale (approx. volume /ml)
  • material: borosilicate glass 3.3
Specifications
Specifications
1000 mL
Borosilicate Glass
2Pack
Polyethylene