Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Conical Separating Funnels with Solid Glass Plug, Scale
Made of DURAN™ borosilicate glass. Lenz Laborglasinstrumente™ Borosilicate Glass Conical Separating Funnels with Solid Glass Plug, Scale are perfect for liquid-liquid extractions and separating chemical components of a mixture into immiscible solvent phases of different densities.
£60.60 - £169.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11822672
|
Lenz Laborglasinstrumente™
04002070 |
1000 mL |
£129.00
Pack of 2 |
Estimated Shipment: 04-12-2024
Log in to see stock. |
|||||
10604491
|
Lenz Laborglasinstrumente™
04002037 |
100 mL |
£67.65
Pack of 2 |
Estimated Shipment: 04-12-2024
Log in to see stock. |
|||||
10664861
|
Lenz Laborglasinstrumente™
04002049 |
250 mL |
£78.80
Pack of 2 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
11836134
|
Lenz Laborglasinstrumente™
04002073 |
2000 mL |
£169.00
Each |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
11812672
|
Lenz Laborglasinstrumente™
04002058 |
500 mL |
£96.95
Pack of 2 |
Estimated Shipment: 04-12-2024
Log in to see stock. |
|||||
11832662
|
Lenz Laborglasinstrumente™
04002028 |
50 mL |
£60.60
Pack of 2 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
Description
- With NS socket and PE stopper
- with solid glass plug
- with screwthread retaining device
- with scale (approx. volume /ml)
- material: borosilicate glass 3.3
Specifications
Borosilicate Glass | |
Polyethylene |