Learn More
BRAND™ Borosilicate Glass Desiccator
Manufactured from borosilicate glass complies with DIN 12491 standards. BRAND™ Borosilicate Glass Desiccator has precisely ground flat flanges. Base ring surface is studded to improve stability and help prevent cracks. Supplied with 24/29 socket and interchangeable borosilicate glass stopcock.
Brand:  BRAND™ 65243
 View more versions of this product
Product Code. 10514262
  • £508.00 / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from borosilicate glass.
  • Complies with DIN 12491 standards.
  • Precisely ground flat flanges.
  • Base ring surface studded to improve stability and help prevent cracks.
  • Interchangeable lid.
  • Supplied with 24/29 socket and interchangeable borosilicate glass stopcock.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
320 mm
Precisely ground flat flanges. Base ring surface studded to improve stability and help prevent cracks.
417 mm
Clear
1
Borosilicate Glass
Socket in the lid with stopcock