Learn More
Lenz Laborglasinstrumente™ Gas Washing Bottles Acc. to Drechsel with Filter Disc
Provides an inexpensive but effective method for the washing and drying of gases. Lenz Laborglasinstrumente™ Gas Washing Bottles Acc. to Drechsel with Filter Disc are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 05510170
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11846084
  • £65.35 / Each
Estimated Shipment: 14-08-2024
to see stock.
Add to basket
Description
Description
  • Acc. to DIN 12596, complete with head
  • Corrosion-free and resistant to thermal shock
  • Available in a range of capacities
Specifications
Specifications
1000 mL
Gas Washing Bottle with Filter Disc
1
DIN 12596
Borosilicate Glass
Filter disc
NS 29/32
1 Pack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products