Learn More
Lenz Laborglasinstrumente™ Gas Washing Bottles Acc. to Drechsel with Filter Disc
Provides an inexpensive but effective method for the washing and drying of gases. Lenz Laborglasinstrumente™ Gas Washing Bottles Acc. to Drechsel with Filter Disc are manufactured from durable DURAN™ borosilicate glass.
Brand: Lenz Laborglasinstrumente™ 05510149
Code : A0
Additional Details : Weight : 0.80000kg
Descripción
- Acc. to DIN 12596, complete with head
- Corrosion-free and resistant to thermal shock
- Available in a range of capacities
Especificaciones
250 mL | |
Gas Washing Bottle with Filter Disc | |
1 | |
DIN 12596 |
Borosilicate Glass | |
Filter disc | |
NS 29/32 | |
1 Pack |