missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ General Purpose Borosilicate Homogenizer Mortars

For use during the initial stages of sample preparation to break down and disrupt a range of materials such as animal or plant tissues, foodstuffs and soil. Fisherbrand™ General Purpose Borosilicate Homogenizer Mortars are tissue grinders manufactured from durable borosilicate glass.

Brand:  Fisherbrand™ 22005F

 View more versions of this product

Product Code. 15371321

  • £120.00 / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Autoclavable
  • Available in a range of capacities
  • For use with Fisherbrand Borosilicate Homogenizer Pestles
Specifications

Specifications

0.152 - 0.254mm
55 mL
Homogenizers
295mm
Autoclavable
Borosilicate Glass
125mm
Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.