missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKA™ Borosilicate Glass Powder Flask
Brand: IKA™ 0003738200
Code : A0
Additional Details : Weight : 0.01000kg
Specifications
Borosilicate Glass | |
RV 10 Rotary Evaporators 500mL | |
500 mL |
Powder Flask | |
1 Each | |
100 g |