Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with PTFE Plug
Made of DURAN™ borosilicate glass. Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with PTFE Plug are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
Brand:  Lenz Laborglasinstrumente™ 04030128
 View more versions of this product
Product Code. 10461711
  • £205.00 / Pack of 2
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With NS socket and PE stopper
  • Without scale, with PTFE plug
  • With screwthread retaining device
  • With inscription label
Specifications
Specifications
50 mL
19/26 NS
2Pack
Borosilicate Glass
2.5mm
Polyethylene