Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with PTFE Plug
Made of DURAN™ borosilicate glass. Lenz Laborglasinstrumente™ Borosilicate Glass Separating Funnels with PTFE Plug are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
£104.00 - £337.00
Product Code | Brand | Capacity (Metric) | Joint Number | Pore Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Joint Number | Pore Size | Price | Quantity & Availability | |||
10102051
|
Lenz Laborglasinstrumente™
04030170 |
1000 mL | 29/32 NS | 6mm |
£337.00
Pack of 2 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||
10373371
|
Lenz Laborglasinstrumente™
04026137 |
100 mL | 29/32 NS | 2.5mm |
£104.00
Pack of 2 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||
10731191
|
Lenz Laborglasinstrumente™
04030137 |
100 mL | 19/26 NS | 2.5mm |
£200.00
Pack of 2 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
12840453
|
Lenz Laborglasinstrumente™
04030149 |
250 mL | 29/32 NS | 4mm |
£166.42
Pack of 2 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||
10694192
|
Lenz Laborglasinstrumente™
04030158 |
500 mL | 29/32 NS | 4mm |
£267.00
Pack of 2 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
10461711
|
Lenz Laborglasinstrumente™
04030128 |
50 mL | 19/26 NS | 2.5mm |
£192.00
Pack of 2 |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||
Description
- With NS socket and PE stopper
- Without scale, with PTFE plug
- With screwthread retaining device
- With inscription label
Specifications
Borosilicate Glass |