Learn More
Kimble Chase™ Borosilicate Glass Shell Vials
Use Kimble™ Borosilicate Glass Shell Vials for storage of dry contents.
Brand:  Kimble Chase™ 60931-12
 View more versions of this product
Product Code. 11873133
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Use Kimble™ Borosilicate Glass Shell Vials for storage of dry contents.
  • Straight-sided
  • Clear N-51A borosilicate glass
  • Without caps
    TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
Borosilicate Glass
35 mm
Yes
2 mL
Ideal for Storing Dry Products
Shell Vial