Learn More
JG Finneran Associates™ Borosilicate Glass Shell Vials
Use these vials for a variety of applications. JG Finneran™ Borosilicate Glass Shell Vials feature thicker walls for safer sample handling.
Brand:  JG Finneran Associates™ 4100-1232
 View more versions of this product
Product Code. 10422751
  • £55.95 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Glass shell vial
  • Available in clear or amber Type I borosilicate glass for easy disposal
  • Limited volume shell vials have a precisely designed conical bottom which permits maximum sample evacuation
Specifications
Specifications
Clear
2 mL
100 Pk.
Borosilicate Glass
100
12 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products