Learn More
JG Finneran Associates™ Borosilicate Glass Shell Vials
Use these vials for a variety of applications. JG Finneran™ Borosilicate Glass Shell Vials feature thicker walls for safer sample handling.
£14.71 - £55.95
Product Code | Brand | Volume (Metric) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Color | Price | Quantity & Availability | ||||
10747651
|
JG Finneran Associates™
4100-840 |
1 mL | Clear |
£14.71
Pack of 100 |
In Stock
Log in to see stock. |
||||
10422751
|
JG Finneran Associates™
4100-1232 |
2 mL | Clear |
£55.95
Pack of 100 |
In Stock
Log in to see stock. |
||||
10625671
|
JG Finneran Associates™
4100-1232A |
2 mL | Amber |
£18.20
Pack of 100 |
In Stock
Log in to see stock. |
||||
Description
- Glass shell vial
- Available in clear or amber Type I borosilicate glass for easy disposal
- Limited volume shell vials have a precisely designed conical bottom which permits maximum sample evacuation
Specifications
Borosilicate Glass | |
100 | |
100 Pk. |