missing translation for 'onlineSavingsMsg'
Learn More

Corning LabGlass Standard Chromatography Column, Fritted

This Standard Chromatography Column is ideal for adsorption, gel, permeation, and ion exchange techniques.

Brand:  Corning 34695-42

Product Code. 15213886

  • £194.00 / Each
Estimated Shipment: 02-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Constructed of durable borosilicate glass.
  • LabGlass Standard Chromatography Glass Columns are made of borosilicate glass and are available with or without TS joints and fritted disks.
  • The TS joints on top of columns permit the connection of a funnel or other reservoir.
  • Fritted disks (40 to 60 μm porosity) support packing materials.
Specifications

Specifications

Adsorption, Gel, Permeation, and Ion Exchange Techniques
9.8 in.
Plain Chromatography Column
250 mm
PTFE Plug to Control Flow Rate
24/40
10.5 mm
40 to 60 μm
Borosilicate Glass
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning LabGlass Standard Chromatography Column, Fritted > Standard Chromatography Column; Dimensions (I.D. x L): 10.5 x 250 mm; Joint Size: 24/40

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.