Learn More
Quickfit™ Borosilicate Glass Stopcock Adapter with Sockets
Control flow rate. QuickFit™ borosilicate glass stopcock adater with cone comes with a GP Rotoflo™ stopcock.
Brand: Quickfit MF14/1/SC
This item is not returnable.
View return policy
Description
- With GP Rotaflo™ PTFE stopcock providing fine flow control and positive closure
- Detachable plastic screwthread connectors allow rubber hose etc. , to be fitted easily and safely
Specifications
Borosilicate Glass | |
14/23 | |
Sockets | |
Stopcock Adapter |