Learn More
Quickfit™ Borosilicate Glass Stopcock Adapter with Sockets
Control flow rate. QuickFit™ borosilicate glass stopcock adater with cone comes with a GP Rotoflo™ stopcock.
£165.00 - £435.00
Specifications
Material | Borosilicate Glass |
---|---|
Includes | Sockets |
Product Type | Stopcock Adapter |
Product Code | Brand | Port Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Port Size | Price | Quantity & Availability | |||||
15557930
|
Quickfit
MF14/1/SC |
14/23 | N/A | ||||||
12415316
|
Quickfit
MF14/2/SC |
19/26 |
£165.00
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||||
15567930
|
Quickfit
MF14/3/SC |
24/29 |
£435.00
Pack of 5 |
Estimated Shipment: 25-02-2025
Log in to see stock. |
|||||
Description
- With GP Rotaflo™ PTFE stopcock providing fine flow control and positive closure
- Detachable plastic screwthread connectors allow rubber hose etc. , to be fitted easily and safely
Specifications
Borosilicate Glass | |
Sockets | |
Stopcock Adapter |