Learn More
Quickfit™ Borosilicate Glass Stopcock Adapter with Sockets
Control flow rate. QuickFit™ borosilicate glass stopcock adater with cone comes with a GP Rotoflo™ stopcock.
Brand: Quickfit MF14/3/SC
431.31 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- With GP Rotaflo™ PTFE stopcock providing fine flow control and positive closure
- Detachable plastic screwthread connectors allow rubber hose etc. , to be fitted easily and safely
Specifications
Borosilicate Glass | |
24/29 | |
Sockets | |
Stopcock Adapter |