Learn More
Fisherbrand™ Borosilicate Glass Tall Form Beaker
Includes graduations and a spout for easy pouring. Fisherbrand™ Borosilicate Glass Tall Form Beaker is available in a variety of sizes.
Brand:  Fisherbrand™ SPFS64030-250
Product Code. 15203199
Description
Description
  • Tall form
  • With spout
  • Graduated
  • Made of borosilicate glass
  • ISO 3819
  • DIN 12331
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Borosilicate Glass Tall Form Beaker > Capacity: 400mL