Learn More
Fisherbrand™ Borosilicate Glass Tall Form Beakers
Includes graduations and a spout for easy pouring. Fisherbrand™ Borosilicate Glass Tall Form Beakers are available in a variety of sizes.
Brand:  Fisherbrand™ SPFS64030-100
 View more versions of this product
Product Code. 15283189
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Borosilicate glass
  • Tall form
  • Graduated
  • With spout
  • Manufactured to ISO 3819, DIN 12331
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Translucent
100 mL
Matte
Spout
Beaker
ISO 3819, DIN 12331
Tall Form
48 mm
Borosilicate Glass
80 mm
10 Pk.
Yes
White Graduated Scale