missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Calibrex™ Universal 520 Bottle-top Dispensers

Constructed with chemically inert contact components for broad chemical resistance, these bottle-top dispensers are universally adapted for most laboratory reagents. Socorex™ Calibrex™ Universal 520 Bottle-top Dispensers are easy to use and maintain.

Brand:  Socorex™ 520.005

 View more versions of this product

Product Code. 10656351

  • £312.00 / Each
Estimated Shipment: 23-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Adjustable volume dispenser.
  • Chemically resistant.
  • Smooth, effortless activation.
  • Base thread fits 32mm bottle neck diameter.
    • Comes with 3 adapters (PP material) for 28, 40 and 45mm bottle neck diameters.
  • Color coding identification.
  • QR coded chemical compatibility
  • Fast, reliable and friendly volume setting:
    • Fast and precise single-hand adjustment.
    • Efficient click-stop mechanism prevents unwanted alteration.
    • Large display is easy to read; the window adjusts to desired side of instrument body.
    • Each step on the cylindrical cam is pre-calibrated and corresponds to one division on the volume graduation.
  • Instrument rotates 360° for adequate positioning on top of the bottle.
  • Air inlet can be enlarged to receive a Luer membrane filter if airborne contamination is a concern.
  • Factory calibrated and easily recalibrated in the laboratory.
  • Disassembly/reassembly without tools.
  • Autoclavable at 121°C / 250°F.
Specifications

Specifications

<±2.0% - <±0.6%
Autoclavable
0.1 mL
Bottle-Calibrex
1 to 5 mL
<0.5% - <0.1%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Socorex™ Calibrex™ Universal 520 Bottle-top Dispensers > Volume: 1-5mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.