Learn More
Sampling Systems™ Bottle Liquid Sampler
Incorporates a spring loaded mechanism, which means that liquid only flows in when the handle is squeezed allowing samples to be taken from set locations. Sampling Systems™ Bottle Liquid Sampler takes samples straight into the bottle.
Brand:  Sampling Systems™ 5031S-1000
 View more versions of this product
Product Code. 15879321
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • 316L Stainless Steel sampler body
  • Glass bottle
  • Single point
  • Sampling for low to medium viscosity liquids
  • Ideal for low volume (unit dose) sampling
  • The sampler is available with a cover that stops the bottle getting wet
Specifications
Specifications
Liquid Sampler
500 mL Glass Bottle
1000 mm
Stainless Steel with Glass Bottle
500 mL